CAS RN 69097-99-0
CAS RN 69097-99-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Hesperetin, MilliporeSigma™ Supelco™
Hesperetin is an aglycon form of flavonoid, which can be found in peels of citrus fruits.
2
phyproof™ Hesperetin, ≥95% (HPLC), MilliporeSigma™ Supelco™
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG