accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6911-87-1

IUPAC Name: 4-bromo-N-methylaniline
Synonyms: 4-bromo-N-methylaniline
Molecular Weight (g/mol): 186.05
Molecular Formula: C7H8BrN
InChi Key: AYVPVDWQZAAZCM-UHFFFAOYSA-N
SMILES: CNC1=CC=C(Br)C=C1
Boiling Point 
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (5)
  • (6)
  • (2)
  • (1)
  • (3)