accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNN

CAS RN 69110-33-4

IUPAC Name: 2,2'-bi1,8-naphthyridine
Synonyms: 2,2'-bi1,8-naphthyridine
Molecular Weight (g/mol): 258.28
Molecular Formula: C16H10N4
InChi Key: OGSULPWGXWPZOJ-UHFFFAOYSA-N
SMILES: C1=CC2=CC=C(N=C2N=C1)C1=NC2=NC=CC=C2C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results