accessibility menu, dialog, popup

UserName

Viewing profile as user:
IN

CAS RN 69113-59-3

IUPAC Name: 3-iodobenzonitrile
Synonyms: 3-iodobenzonitrile
Molecular Weight (g/mol): 229.02
Molecular Formula: C7H4IN
InChi Key: BGARPMGQRREXLN-UHFFFAOYSA-N
SMILES: IC1=CC=CC(=C1)C#N
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)