accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOH3C

CAS RN 69169-71-7

IUPAC Name: 8-methoxy-4-methyl-2H-benzo[g]chromen-2-one
Synonyms: 8-methoxy-4-methylbenzo[g]chromen-2-one
Molecular Weight (g/mol): 240.26
Molecular Formula: C15H12O3
InChi Key: GWFUYESISQUIHB-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=C3C(C)=CC(=O)OC3=CC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results