CAS RN 692-86-4
CAS RN 692-86-4
IUPAC Name:
ethyl undec-10-enoate
Synonyms:
ethyl undecylenate
Molecular Weight (g/mol):
212.33
Molecular Formula:
C13H24O2
InChi Key:
FXNFFCMITPHEIT-UHFFFAOYSA-N
SMILES:
CCOC(=O)CCCCCCCCC=C
1
–
1
of
1
results
1
Ethyl 10-Undecenoate 97.0+%, TCI America™

CAS: 692-86-4 Molecular Formula: C13H24O2 Molecular Weight (g/mol): 212.333 MDL Number: MFCD00009220 InChI Key: FXNFFCMITPHEIT-UHFFFAOYSA-N Synonym: 10-Undecenoic Acid Ethyl Ester PubChem CID: 12729 IUPAC Name: ethyl undec-10-enoate SMILES: CCOC(=O)CCCCCCCCC=C