accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6920-24-7

IUPAC Name: hexadecane-1,2-diol
Synonyms: cetyl glycol
Molecular Weight (g/mol): 258.45
Molecular Formula: C16H34O2
InChi Key: BTOOAFQCTJZDRC-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCC(O)CO
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results