accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6921-34-2

IUPAC Name: benzyl(chloro)magnesium
Synonyms: benzyl(chloro)magnesium
Molecular Weight (g/mol): 150.89
Molecular Formula: C7H7ClMg
InChi Key: NRAFPLGJPPJUNB-UHFFFAOYSA-M
SMILES: Cl[Mg]CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)