accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CO

CAS RN 6921-35-3

IUPAC Name: 3,3-dimethyloxetane
Synonyms: oxetane, 3,3-dimethyl-
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: RVGLUKRYMXEQAH-UHFFFAOYSA-N
SMILES: CC1(C)COC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results