CAS RN 69227-47-0
IUPAC Name:
tris(3,5-dimethylphenyl)phosphane
Synonyms:
tris(3,5-dimethylphenyl)phosphane
Molecular Weight (g/mol):
346.45
Molecular Formula:
C24H27P
InChi Key:
XRALRSQLQXKXKP-UHFFFAOYSA-N
SMILES:
CC1=CC(=CC(C)=C1)P(C1=CC(C)=CC(C)=C1)C1=CC(C)=CC(C)=C1
1
–
1
of
1
results
1
Sigma Aldrich (-)-Butyl L-Lactate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.