accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6924-66-9

IUPAC Name: quinoxaline-5-carboxylic acid
Synonyms: quinoxaline-5-carboxylic acid
Molecular Weight (g/mol): 174.16
Molecular Formula: C9H6N2O2
InChi Key: QLZNISOPACYKOR-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2N=CC=NC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results