accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClHHONCH2N

CAS RN 69257-04-1

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 421.99
Molecular Formula: C26H30ClN2O
InChi Key: NNUGFLHUZJYXIK-UHFFFAOYNA-N
SMILES: [H].[Cl].OC(C1CC2CC[N+]1(CC1=CC=CC=C1)CC2C=C)C1=C2C=CC=CC2=NC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results