accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 69261-01-4

IUPAC Name: 4-fluoro-N,N-dimethyl-2-nitroaniline
Synonyms: 4-fluoro-N,N-dimethyl-2-nitroaniline
Molecular Weight (g/mol): 184.17
Molecular Formula: C8H9FN2O2
InChi Key: RGCUVPLVTHHXMT-UHFFFAOYSA-N
SMILES: CN(C)C1=C(C=C(F)C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results