accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6929-08-4

IUPAC Name: undecan-3-ol
Synonyms: undecan-3-ol
Molecular Weight (g/mol): 172.31
Molecular Formula: C11H24O
InChi Key: HCARCYFXWDRVBZ-UHFFFAOYNA-N
SMILES: CCCCCCCCC(O)CC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results