accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSCH3

CAS RN 693-83-4

IUPAC Name: 1-(decylsulfanyl)decane
Synonyms: 1-(decylsulfanyl)decane
Molecular Weight (g/mol): 314.62
Molecular Formula: C20H42S
InChi Key: RKYMVQJWYYOIJB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCSCCCCCCCCCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)