accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 69304-47-8

IUPAC Name: 5-[(1E)-2-bromoethenyl]-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: brivudine
Molecular Weight (g/mol): 333.14
Molecular Formula: C11H13BrN2O5
InChi Key: ODZBBRURCPAEIQ-PIXDULNESA-N
SMILES: OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(\C=C\Br)C(=O)NC1=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)