accessibility menu, dialog, popup

UserName

Viewing profile as user:
HCl(S)(S)ONH2OCH3H3CCH3CH3CH3

CAS RN 69320-89-4

IUPAC Name: tert-butyl (2S,3S)-2-amino-3-methylpentanoate hydrochloride
Synonyms: tert-butyl (2S,3S)-2-amino-3-methylpentanoate hydrochloride
Molecular Weight (g/mol): 223.74
Molecular Formula: C10H22ClNO2
InChi Key: IFRYMHOZFAPYPJ-HAGKNZRZNA-N
SMILES: Cl.CC[C@H](C)[C@H](N)C(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)