accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrBr

CAS RN 69321-60-4

IUPAC Name: 1,3-dibromo-2-methylbenzene
Synonyms: 1,3-dibromo-2-methylbenzene
Molecular Weight (g/mol): 249.93
Molecular Formula: C7H6Br2
InChi Key: OCSKCBIGEMSDIS-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=CC=C1Br
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results