CAS RN 69355-99-3
IUPAC Name:
(2R,3S)-3-(benzyloxy)-2-{[(tert-butoxy)carbonyl]amino}butanoic acid
Synonyms:
(2R,3S)-3-(benzyloxy)-2-[(tert-butoxycarbonyl)amino]butanoic acid|BOC-D-THR(BZL)-OH
Molecular Weight (g/mol):
309.36
Molecular Formula:
C16H23NO5
InChi Key:
CTXPLTPDOISPTE-VEZULTJBNA-N
SMILES:
C[C@H](OCC1=CC=CC=C1)[C@@H](NC(=O)OC(C)(C)C)C(O)=O
1
–
1
of
1
results
1
N-Boc-O-benzyl-D-threonine, 98%, Thermo Scientific™
CAS: 69355-99-3 Molecular Formula: C16H23NO5 Molecular Weight (g/mol): 309.36 MDL Number: MFCD00038247 InChI Key: CTXPLTPDOISPTE-VEZULTJBNA-N Synonym: boc-d-thr bzl-oh,boc-o-benzyl-d-threonine,2r,3s-3-benzyloxy-2-tert-butoxycarbonyl amino butanoic acid,ambotzbaa1378,boc-d-thr obzl,pubchem12446,n-boc-d-thr bzl-oh,boc-d-thr bzl-oh tlc,o-benzyl-n-tert-butyloxycarbonyl-d-threonine PubChem CID: 6992573 SMILES: C[C@H](OCC1=CC=CC=C1)[C@@H](NC(=O)OC(C)(C)C)C(O)=O