CAS RN 6936-48-7
CAS RN 6936-48-7
IUPAC Name:
(2Z)-3-(methylcarbamoyl)prop-2-enoic acid
Synonyms:
(2Z)-3-(methylcarbamoyl)prop-2-enoic acid
Molecular Weight (g/mol):
129.12
Molecular Formula:
C5H7NO3
InChi Key:
DFQUBYCHLQAFOW-IHWYPQMZSA-N
SMILES:
CNC(=O)\C=C/C(O)=O
1
–
1
of
1
results
1
N-Methylmaleic Acid Monoamide 98.0+%, TCI America™
CAS: 6936-48-7 Molecular Formula: C5H7NO3 Molecular Weight (g/mol): 129.115 MDL Number: MFCD00014012 InChI Key: DFQUBYCHLQAFOW-IHWYPQMZSA-N Synonym: n-methylmaleamic acid,z-4-methylamino-4-oxobut-2-enoic acid,n-methylmaleic acid monoamide,2z-3-methylcarbamoyl prop-2-enoic acid,maleamic acid, n-methyl,2z-4-methylamino-4-oxo-2-butenoic acid,3-methylaminocarbonyl acrylic acid,n-methylmaleamic,hifl@nafr vvfzafh,n-methyl maleamic acid PubChem CID: 5354909 IUPAC Name: (Z)-4-(methylamino)-4-oxobut-2-enoic acid SMILES: CNC(=O)C=CC(=O)O