CAS RN 69373-37-1
IUPAC Name:
7-nitro-3,4-dihydro-2H-1,4-benzothiazine
Synonyms:
7-nitro-3,4-dihydro-2H-1,4-benzothiazine
Molecular Weight (g/mol):
196.22
Molecular Formula:
C8H8N2O2S
InChi Key:
LCOAYUPAESYAHF-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC2=C(NCCS2)C=C1
1
–
1
of
1
results
1
Promotions Available
7-Nitro-3,4-dihydro-2H-1,4-benzothiazine, 97%, Thermo Scientific Chemicals
7-Nitro-3,4-dihydro-2H-1,4-benzothiazine, 97%, Thermo Scientific Chemicals
CAS: 69373-37-1 Molecular Formula: C8H8N2O2S Molecular Weight (g/mol): 196.22 MDL Number: MFCD23379852 InChI Key: LCOAYUPAESYAHF-UHFFFAOYSA-N Synonym: 7-nitro-3,4-dihydro-2h-benzo b 1,4 thiazine PubChem CID: 20496121 IUPAC Name: 7-nitro-3,4-dihydro-2H-1,4-benzothiazine SMILES: [O-][N+](=O)C1=CC2=C(NCCS2)C=C1