accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2ClFNOOHOCl

CAS RN 69377-81-7

IUPAC Name: 2-[(4-amino-3,5-dichloro-6-fluoropyridin-2-yl)oxy]acetic acid
Synonyms: fluroxypyr
Molecular Weight (g/mol): 255.03
Molecular Formula: C7H5Cl2FN2O3
InChi Key: MEFQWPUMEMWTJP-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C(F)=NC(OCC(O)=O)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results