accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CBr

CAS RN 6938-66-5

IUPAC Name: 1-bromodocosane
Synonyms: 1-bromodocosane
Molecular Weight (g/mol): 389.51
Molecular Formula: C22H45Br
InChi Key: QYOXLKAKUAASNA-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCBr
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results