accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOOOCH3CH3

CAS RN 6938-94-9

IUPAC Name: 1,6-bis(propan-2-yl) hexanedioate
Synonyms: diisopropyl adipate wickenol 116 crodamol DA standamul dipa prodipate tegester 504-D β dia schercemol dia ceraphyl 230 bis(1-methylethyl)hexanedioate
Molecular Weight (g/mol): 230.3
Molecular Formula: C12H22O4
InChi Key: ZDQWESQEGGJUCH-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)CCCCC(=O)OC(C)C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)