accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOOBrCH3

CAS RN 69383-59-1

IUPAC Name: 1-bromo-2,4-dimethyl-5-nitrobenzene
Synonyms: 1-bromo-2,4-dimethyl-5-nitrobenzene
Molecular Weight (g/mol): 230.06
Molecular Formula: C8H8BrNO2
InChi Key: HYYJANCKMGXHLQ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(Br)C=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results