accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3H3CCH3HNOHO

CAS RN 6939-23-7

IUPAC Name: hydrogen 2-(tert-butylamino)acetic acid chloride
Synonyms: hydrogen T-butylglycine chloride
Molecular Weight (g/mol): 167.63
Molecular Formula: C6H14ClNO2
InChi Key: QORBVLNEGUIPND-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC(C)(C)NCC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)