accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6939-93-1

IUPAC Name: 4-bromobenzene-1,3-dicarboxylic acid
Synonyms: 4-bromobenzene-1,3-dicarboxylic acid
Molecular Weight (g/mol): 245.03
Molecular Formula: C8H5BrO4
InChi Key: MSQIEZXCNYUWHN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(Br)C(=C1)C(O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)