CAS RN 694-31-5
IUPAC Name:
1,5-dimethyl-1H-pyrazole
Synonyms:
1,5-dimethylpyrazole
Molecular Weight (g/mol):
96.13
Molecular Formula:
C5H8N2
InChi Key:
LSZQMSSIUQNTDX-UHFFFAOYSA-N
SMILES:
CN1N=CC=C1C
1
–
1
of
1
results
1
1,5-Dimethylpyrazole 98.0+%, TCI America™
CAS: 694-31-5 Molecular Formula: C5H8N2 Molecular Weight (g/mol): 96.13 MDL Number: MFCD01646129 InChI Key: LSZQMSSIUQNTDX-UHFFFAOYSA-N PubChem CID: 136502 IUPAC Name: 1,5-dimethyl-1H-pyrazole SMILES: CN1N=CC=C1C