accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 694-82-6

IUPAC Name: 2-fluorocyclohexan-1-one
Synonyms: 2-fluorocyclohexan-1-one
Molecular Weight (g/mol): 116.14
Molecular Formula: C6H9FO
InChi Key: VQYOFTVCYSPHPG-UHFFFAOYNA-N
SMILES: FC1CCCCC1=O
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)