accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NO

CAS RN 694-85-9

IUPAC Name: 1-methyl-1,2-dihydropyridin-2-one
Synonyms: 2(1H)-pyridinone, 1-methyl-
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: DVVGIUUJYPYENY-UHFFFAOYSA-N
SMILES: CN1C=CC=CC1=O
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)