accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6940-53-0

IUPAC Name: 1-chloro-2,5-dimethoxy-4-nitrobenzene
Synonyms: 1-chloro-2,5-dimethoxy-4-nitrobenzene
Molecular Weight (g/mol): 217.61
Molecular Formula: C8H8ClNO4
InChi Key: ORLPGMKKCAEWOW-UHFFFAOYSA-N
SMILES: COC1=CC(=C(OC)C=C1Cl)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results