CAS RN 6940-58-5
CAS RN 6940-58-5
IUPAC Name:
pentane-1,3,5-tricarboxylic acid
Synonyms:
pentane-1,3,5-tricarboxylic acid
Molecular Weight (g/mol):
204.18
Molecular Formula:
C8H12O6
InChi Key:
ROTJZTYLACIJIG-UHFFFAOYSA-N
SMILES:
OC(=O)CCC(CCC(O)=O)C(O)=O
1
–
1
of
1
results
1
1,3,5-Pentanetricarboxylic Acid 98.0+%, TCI America™

CAS: 6940-58-5 Molecular Formula: C8H12O6 Molecular Weight (g/mol): 204.18 MDL Number: MFCD00020546 InChI Key: ROTJZTYLACIJIG-UHFFFAOYSA-N PubChem CID: 96220 IUPAC Name: pentane-1,3,5-tricarboxylic acid SMILES: OC(=O)CCC(CCC(O)=O)C(O)=O