CAS RN 69403-12-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MP Biomedicals, Inc L-Phenylalanine dehydrogenase, MP Biomedicals™
CAS: 69403-12-9 Synonym: E.C. 1.4.1.20