accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 69409-98-9

IUPAC Name: 2-fluoro-4-(trifluoromethyl)aniline
Synonyms: 2-fluoro-4-(trifluoromethyl)aniline
Molecular Weight (g/mol): 179.12
Molecular Formula: C7H5F4N
InChi Key: ARHDUOQIXLGANT-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results