accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6941-37-3

IUPAC Name: hexadecyltrimethylazanium perchlorate
Synonyms: cetyl-trimethyl-ammonium perchlorate
Molecular Weight (g/mol): 384
Molecular Formula: C19H42ClNO4
InChi Key: KXNZYPBRSATHKV-UHFFFAOYSA-M
SMILES: [O-][Cl](=O)(=O)=O.CCCCCCCCCCCCCCCC[N+](C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results