accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 69411-67-2

IUPAC Name: 2-fluoro-4-nitro-1-(trifluoromethyl)benzene
Synonyms: 2-fluoro-4-nitro-1-(trifluoromethyl)benzene
Molecular Weight (g/mol): 209.1
Molecular Formula: C7H3F4NO2
InChi Key: XYKVQPNHGFUXBS-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results