accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3BrH3CBr

CAS RN 6942-99-0

IUPAC Name: 2,4-dibromo-1,3,5-trimethylbenzene
Synonyms: 2,4-dibromo-1,3,5-trimethylbenzene
Molecular Weight (g/mol): 277.99
Molecular Formula: C9H10Br2
InChi Key: CIHJFEWFZJQTFE-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(Br)C(C)=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results