CAS RN 69430-36-0
IUPAC Name:
2-bromo-2-chloroacetic acid
Synonyms:
bromochloroacetic acid keratin
Molecular Weight (g/mol):
173.39
Molecular Formula:
C2H2BrClO2
InChi Key:
GEHJBWKLJVFKPS-UHFFFAOYNA-N
SMILES:
OC(=O)C(Cl)Br
1
–
1
of
1
results
1
Promotions Available
Keratin, Powder, Spectrum™ Chemical
Keratin, Powder, Spectrum™ Chemical
CAS: 69430-36-0 Molecular Formula: C2H2BrClO2 Molecular Weight (g/mol): 173.39 InChI Key: GEHJBWKLJVFKPS-UHFFFAOYNA-N SMILES: OC(=O)C(Cl)Br