accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NBrNOO

CAS RN 6945-68-2

IUPAC Name: 5-bromo-3-nitropyridin-2-amine
Synonyms: 5-bromo-3-nitropyridin-2-amine
Molecular Weight (g/mol): 218.01
Molecular Formula: C5H4BrN3O2
InChi Key: QOOCOFOGYRQPPN-UHFFFAOYSA-N
SMILES: NC1=NC=C(Br)C=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)