accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3COONHNH2

CAS RN 6945-92-2

IUPAC Name: hydrogen ethyl 2-hydrazinylacetate chloride
Synonyms: hydrogen ethyl 2-hydrazinylacetate chloride
Molecular Weight (g/mol): 154.59
Molecular Formula: C4H11ClN2O2
InChi Key: HZZRIIPYFPIKHR-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCOC(=O)CNN
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)