accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNOOF

CAS RN 694501-34-3

IUPAC Name: 1-ethyl-4-(2-fluoro-4-nitrophenyl)piperazine
Synonyms: 1-ethyl-4-(2-fluoro-4-nitrophenyl)piperazine
Molecular Weight (g/mol): 253.28
Molecular Formula: C12H16FN3O2
InChi Key: XMJXGMZBNFICRS-UHFFFAOYSA-N
SMILES: CCN1CCN(CC1)C1=C(F)C=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results