accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6949-73-1

IUPAC Name: 2-hydroxy-9H-fluoren-9-one
Synonyms: 2-hydroxy-9-fluorenone
Molecular Weight (g/mol): 196.21
Molecular Formula: C13H8O2
InChi Key: GXUBPHMYNSICJC-UHFFFAOYSA-N
SMILES: OC1=CC=C2C3=CC=CC=C3C(=O)C2=C1
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)