accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNH2

CAS RN 69491-57-2

IUPAC Name: 4-(pyrimidin-2-yl)aniline
Synonyms: 4-(pyrimidin-2-yl)aniline
Molecular Weight (g/mol): 171.2
Molecular Formula: C10H9N3
InChi Key: RUVNVJCKWOOLOH-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C1=NC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results