accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClBr

CAS RN 695-96-5

IUPAC Name: 2-bromo-4-chlorophenol
Synonyms: 2-bromo-4-chlorophenol
Molecular Weight (g/mol): 207.45
Molecular Formula: C6H4BrClO
InChi Key: ZIYRDJLAJYTELF-UHFFFAOYSA-N
SMILES: OC1=CC=C(Cl)C=C1Br
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)