accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3H3C

CAS RN 695-98-7

IUPAC Name: 2,3,5-trimethylpyridine
Synonyms: pyridine, 2,3,5-trimethyl-
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: GFYHSKONPJXCDE-UHFFFAOYSA-N
SMILES: CC1=CN=C(C)C(C)=C1
Boiling Point 
  • (1)
  • (3)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)