accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNO

CAS RN 6953-22-6

IUPAC Name: 5-(benzyloxy)-1H-indole-3-carbaldehyde
Synonyms: 5-(benzyloxy)-1H-indole-3-carbaldehyde
Molecular Weight (g/mol): 251.29
Molecular Formula: C16H13NO2
InChi Key: DJGNUBADRQIDNQ-UHFFFAOYSA-N
SMILES: O=CC1=CNC2=CC=C(OCC3=CC=CC=C3)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results