accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6953-61-3

IUPAC Name: N-hydroxynaphthalene-1-carboxamide
Synonyms: N-hydroxynaphthalene-1-carboxamide
Molecular Weight (g/mol): 187.2
Molecular Formula: C11H9NO2
InChi Key: JRZGPWOEHDOVMC-UHFFFAOYSA-N
SMILES: ONC(=O)C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results