accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 69556-70-3

IUPAC Name: 4-methoxy-2,5-dihydrofuran-2-one
Synonyms: 4-methoxy-5H-furan-2-one
Molecular Weight (g/mol): 114.1
Molecular Formula: C5H6O3
InChi Key: VOYDEHILKLSVNN-UHFFFAOYSA-N
SMILES: COC1=CC(=O)OC1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)