CAS RN 695816-47-8
IUPAC Name:
1-[2-(1,3-dioxolan-2-yl)phenyl]-2,5-dimethylphospholane
Synonyms:
1-[2-(1,3-dioxolan-2-yl)phenyl]-2,5-dimethylphospholane
Molecular Weight (g/mol):
264.31
Molecular Formula:
C15H21O2P
InChi Key:
JAKXTLMNIHPLEU-UHFFFAOYNA-N
SMILES:
CC1CCC(C)P1C1=CC=CC=C1C1OCCO1
1
–
1
of
1
results
1
Promotions Available
(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-dimethylphospholane, 97%, Thermo Scientific™
(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-dimethylphospholane, 97%, Thermo Scientific™
CAS: 695816-47-8 Molecular Formula: C15H21O2P Molecular Weight (g/mol): 264.3 InChI Key: JAKXTLMNIHPLEU-RYUDHWBXSA-N Synonym: 2s,5s-+-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane,2s,5s-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane,s,s-me-rajphos,2-2-2s,5s-2,5-dimethylphospholan-1-yl phenyl-1,3-dioxolane,2s,5s-+-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, kanata purity,1-2-1,3-dioxolane-2-yl phenyl-2beta,5alpha-dimethyltetrahydro-1h-phosphole,2-2-2s,5s-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, min PubChem CID: 16218307 IUPAC Name: 2-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]phenyl]-1,3-dioxolane SMILES: CC1CCC(P1C2=CC=CC=C2C3OCCO3)C