accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 696-02-6

IUPAC Name: 4-chloro-1,2-difluorobenzene
Synonyms: 4-chloro-1,2-difluorobenzene
Molecular Weight (g/mol): 148.54
Molecular Formula: C6H3ClF2
InChi Key: OPQMRQYYRSTBME-UHFFFAOYSA-N
SMILES: FC1=CC=C(Cl)C=C1F
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)